Anti-GABRB3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9687
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor beta 3
- Antigen symbol:GABRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:24922
- Antigen synonyms:beta3|Cp1|Gamma aminobutyric acid A receptor beta 3,A230092K12Rik|AW049585|Gabrb-3
- Storage buffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to rat Gabrb3.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against a recombinant Gabrb3.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat