Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ANP32C Rabbit Polyclonal Antibody
Anti-ANP32C Rabbit Polyclonal Antibody
Catalog # ABNOPAB17337
Supplier:  Abnova
Anti-ANP32C Rabbit Polyclonal Antibody
Catalog # ABNOPAB17337
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acidic Nuclear Phosphoprotein 32 Family, Member C
  • Antigen symbol:
    ANP32C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    23520
  • Antigen synonyms:
    Tumorigenic protein pp32r1|Phosphoprotein 32-related protein 1|PP32R1
  • Storage buffer:
    PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to internal of human ANP32C.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of ANP32C.

Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:10000

Type: Primary
Antigen: ANP32C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human