Anti-FAS Rabbit Polyclonal Antibody
Catalog # ABNOPAB8028
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Antigen symbol:FAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:246097
- Antigen synonyms:FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
- Storage buffer:0,9 mg NaCl, 0,2 mg Na2HPO4 (5 mg BSA, 0,05 mg sodium azide, 0,05 mg Thimerosal)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 200 ?l of deionized water and concentration will be 500 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at N-terminus of rat Fas.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Fas.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml; Immunohistochemistry (Frozen sections): 1-2 ?g/ml
Type: Primary
Antigen: FAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat