Anti-FFAR2 Rabbit Polyclonal Antibody
ABNOPAB26348
: Abnova
- Antibody type:Primary
- Antigen name:Free fatty acid receptor 2
- Antigen symbol:FFAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Gorilla,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2867
- Antigen synonyms:FFA2R|GPR43
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 17 amino acids at internal region of human FFAR2.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of FFAR2.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml
Type: Primary
Antigen: FFAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human