Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-GABBR1 Rabbit Polyclonal Antibody
  ABNOPAB26218
 :  Abnova
undefined
Anti-GABBR1 Rabbit Polyclonal Antibody
  ABNOPAB26218
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Gamma-Aminobutyric Acid (GABA) B Receptor, 1
  • Antigen symbol:
    GABBR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Hamster,
    Pig,
    Rabbit,
    Monkey,
    Gorilla,
    Orangutan,
    Human,
    Bovine,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2550
  • Antigen synonyms:
    GABABR1|GABBR1-3|GPRC3A|GB1|dJ271M21.1.2|dJ271M21.1.1
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 15 amino acid at N-terminus of human GABBR1.
  • Purification:
    Immunoaffinity chromatography
  • Size:
    50 μg
  • Pk:
    50 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of GABBR1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 9-18 ?g/ml

Type: Primary
Antigen: GABBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Gorilla, Hamster, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat