Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HCAR3 Rabbit Polyclonal Antibody
Anti-HCAR3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26279
Supplier:  Abnova
Anti-HCAR3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26279
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    hydroxycarboxylic acid receptor 3
  • Antigen symbol:
    HCAR3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Hamster,
    Gorilla,
    Human,
    Dog
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8843
  • Antigen synonyms:
    HM74|GPR109B|HCA3|Puma-g|PUMAG
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 20 amino acids at 3rd cytoplasmic domain of human GPR109B.
  • Purification:
    Immunoaffinity chromatography
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of GPR109B.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 19-38 ?g/ml

Type: Primary
Antigen: HCAR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Hamster, Human