Anti-MIR7-3HG Rabbit Polyclonal Antibody
ABGEAP18854C
: Abgent
- Antibody type:Primary
- Antigen symbol:MIR7-3HG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Amino acid number:26 to 54
- Storage buffer:pH 7,4
- Molecular weight:14.321 kDa
- Sequence:PNIRTAPLGRSEKRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.65
- Shipping temperature:4 °C
- Immunogen:This PGSF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 26-54 amino acids from the Central region of human PGSF1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: MIR7-3HG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human