Anti-iPLA2 Rabbit Polyclonal Antibody
CAYM160507-1
- Antibody type:Primary
- Antigen symbol:IPLA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Hamster,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized
- Amino acid number:557 to 576
- Sequence:PRFNQNINLKPPTQPADQLV
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:hamster iPLA2 amino acids 557-576 (PRFNQNINLKPPTQPADQLV)
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: iPLA2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Hamster, Human, Mouse, Rat