Anti-COX-2 Mouse Monoclonal Antibody [clone: CX229]
CAYM160112-1
- Antibody type:Primary
- Antigen symbol:COX-2
- Clonality:Monoclonal
- Clone:CX229
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Monkey,Sheep,Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized
- Amino acid number:580 to 599
- Sequence:NASSSRSGLDDINPTVLLKE
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human COX-2 amino acids 580-599
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: COX-2
Clonality: Monoclonal
Clone: CX229
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Sheep