Anti-JARID1B/PLU1 Rabbit Polyclonal Antibody
Catalog # CAYM14701-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:JARID1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:No
- Antigen synonyms:PLU1
- Amino acid number:1534 to 1544
- Storage buffer:TBS, pH 7,4, containing 50% glycerol, 0,1% BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human JARID1B/PLU1 amino acids 1,534-1,544
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: JARID1B/PLU1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human