Anti-GPR31 Rabbit Polyclonal Antibody
Catalog # CAYM14083-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:GPR31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:288 to 298
- Storage buffer:TBS, pH 7,4
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human GPR31 amino acids 288-298
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: GPR31
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human