Anti-TXA2-R Rabbit Polyclonal Antibody
Catalog # CAYM10004452-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:TXA2-R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse,Cos-7
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:No
- Form:lyophilized
- Amino acid number:323 to 343
- Storage buffer:TBS, pH 7,4
- Sequence:LSTRPRSLSLQPQLTQRSGLQ
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human TP receptor C-terminal amino acids 323-343 (LSTRPRSLSLQPQLTQRSGLQ)
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: TXA2-R
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: cos-7, Human, Mouse, Rat