Anti-6xHis Mouse Monoclonal Antibody (DyLight® 800) [clone: 33D10.D2]
ROCK200-345-382
- Antibody type:Primary
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:33D10.D2
- Conjugation:DyLight® 800
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D10.D2
Conjugation: DyLight® 800
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: