Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Cyclin D1 Mouse Monoclonal Antibody [clone: DCS-6]
Anti-Cyclin D1 Mouse Monoclonal Antibody [clone: DCS-6]
Catalog # USBI138535-100
Supplier:  US Biological
Anti-Cyclin D1 Mouse Monoclonal Antibody [clone: DCS-6]
Catalog # USBI138535-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin D1
  • Antigen symbol:
    CCND1
  • Clonality:
    Monoclonal
  • Clone:
    DCS-6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    full-length
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~0,2 mg/ml
  • Immunogen:
    Recombinant protein corresponding to full length human cyclin D1.
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Cyclin D1
Clonality: Monoclonal
Clone: DCS-6
Conjugation: unconjugated
Epitope: full-length
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Monkey, Mouse, Rat,