Anti-M13, fd, F1 Bacteriophages, Filamentous Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 5F167]
Catalog # USBIM1011-50D-250
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:M13, fd, F1 Bacteriophages, Filamentous
- Antigen symbol:M13
- Clonality:Monoclonal
- Clone:5F167
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- Isotype:IgG2b
- Reactivity:E. coli
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:Tris, pH 8.0, 0,5% BSA, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,4 mg/ml
- Immunogen:fd phages from E. coli F+ strain (JM109).
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:250 μl
- Pk:250 µl
Specifications
About this item
Recommended dilution: ELISA: 1 ?g/ml
Type: Primary
Antigen: M13, fd, F1 Bacteriophages, Filamentous
Clonality: Monoclonal
Clone: 5F167
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: E.coli,