Anti-Transferrin Rabbit Polyclonal Antibody
Catalog # USBIT8199-04K-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Transferrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:–20 °C
- Immunogen:Purified rat serum Transferrin (Tf).
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000-1:10000; Western Blot: 1:1000-1:5000
Type: Primary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat,