Anti-Angiotensin Converting Enzyme 2 Mouse Monoclonal Antibody [clone: 7C9]
Catalog # USBIA2295-01W9-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Angiotensin Converting Enzyme 2
- Antigen symbol:ACE2
- Clonality:Monoclonal
- Clone:7C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Amino acid number:18 to 740
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Purified NSO-derived recombinant human angiotensin I converting enzyme-2 (rhACE-2) ectodomain (aa18-740).
- Purification:purified by Protein G chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:10K-1:30K; Western Blot: 1-2?g/ml
Type: Primary
Antigen: Angiotensin Converting Enzyme 2
Clonality: Monoclonal
Clone: 7C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,