Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 1A12]
ORIGTA507387
: OriGene
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 7
- Antigen symbol:SIGLEC7
- Clonality:Monoclonal
- Clone:1A12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:41.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SIGLEC7(NP_057627) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human