Anti-TUBB Mouse Monoclonal Antibody [clone: 87CT59.3.7]
Catalog # ORIGTA324326
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TUBB
- Clonality:Monoclonal
- Clone:87CT59.3.7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Hamster,Pig,Xenopus,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Purified monoclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:49.67 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:TBB5 recombinant protein is used to produce this monoclonal antibody.
- Purification:purified by Protein G chromatography, followed by dialysis against PBS
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Immunofluorescence: 1:25; Immunochemistry: 1:10-50; Western Blot: 1:100-500
Type: Primary
Antigen: TUBB
Clonality: Monoclonal
Clone: 87CT59.3.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Chicken, Hamster, Human, Mouse, Pig, Rat, Xenopus