Anti-RASSF1 Mouse Polyclonal Antibody [clone: eB114-10H1]
ORIGTA320455
: OriGene
- Antibody type:Primary
- Antigen name:Ras association (RalGDS/AF-6) domain family member 1
- Antigen symbol:RASSF1
- Clonality:Polyclonal
- Clone:eB114-10H1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:25 μg
- Pk:25 µG
Type: Primary
Antigen: RASSF1
Clonality: Polyclonal
Clone: eB114-10H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human