Anti-CD70 Rat Polyclonal Antibody [clone: FR70]
ORIGTA320290
: OriGene
- Antibody type:Primary
- Antigen symbol:CD70
- Clonality:Polyclonal
- Clone:FR70
- Conjugation:Unconjugated
- Host:Rat
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: CD70
Clonality: Polyclonal
Clone: FR70
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse