Anti-RARA Mouse Monoclonal Antibody [clone: 1C6]
ORIGCF806338
: OriGene
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Alpha
- Antigen symbol:RARA
- Clonality:Monoclonal
- Clone:1C6
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:52 to 299
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 52-299 of human RARA(NP_000955) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: RARA
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human