Anti-TAB3 Rabbit Polyclonal Antibody
ORIGTA320155
: OriGene
- Antibody type:Primary
- Antigen name:TGF-beta activated kinase 3
- Antigen symbol:TAB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:TAB3 Antibody is supplied in PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:TAB3 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of human TAB3.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: TAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human