Anti-BrdU Monoclonal Antibody [clone: 2B5]
Catalog # ORIGTA190219
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:BRDU
- Clonality:Monoclonal
- Clone:2B5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Bromodeoxyuridine coupled to keyhole limpet hemocyanin (KLH).
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150; Immunofluorescence: 1:150
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: