Anti-RARRES1 Mouse Monoclonal Antibody [clone: 1A7]
Catalog # ORIGCF506090
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Retinoic acid receptor responder (tazarotene induced) 1
- Antigen symbol:RARRES1
- Clonality:Monoclonal
- Clone:1A7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:33.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human RARRES1(NP_996846) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: RARRES1
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human