Anti-SIGLEC9 Mouse Monoclonal Antibody [clone: 8F7]
Catalog # ORIGCF500380
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 9
- Antigen symbol:SIGLEC9
- Clonality:Monoclonal
- Clone:8F7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Dog
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:50.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SIGLEC9 (NP_055256) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2500-5000; Immunochemistry: 1:50
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone: 8F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey