Chemical preservative vials, Vialservatives™
These Vialservatives™ make proper preservation at the point of sample collection convenient and safer than handling glass ampoules or larger volume chemical bottles.
- Allows addition of preservative immediately after sample collection
- Threaded vial containing a premeasured aliquot of preservative chemical solution
- Individually labeled with contents and colour coded labels to minimize error in the field
Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification; manufactured under an ISO certified quality management system.
- Catalog No:
- EPSCSVCN-1
- EPSCSVCN-2-1
- EPSCSVCN-2
- EPSCSVCS-1-1
- EPSCSVCN-5
- EPSCSVCS-1-3
- EPSCSVCH-5-1
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together