Anti-Rab9 Rabbit Antibody
BSBTPA2281
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-9
- Antigen symbol:RAB9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:178 - 193
- Sequence:RSDHLIQTDTVNLHRK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Rab9 (178-193aa RSDHLIQTDTVNLHRK), identical to the related rat sequence, and different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ras-related protein Rab-9A(RAB9A) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: RAB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat