Order Entry
United Kingdom
ContactUsLinkComponent
Conradson test unit
Conradson test unit
Catalog # STAN10610-0
Supplier:  STANHOPE SETA
undefined
Conradson test unit
Catalog # STAN10610-0
Supplier:  STANHOPE SETA

Specifications

  • Description:
    Conradson test unit
  • W×D×H:
    230×180×180 mm
  • Weight:
    2100 g
  • Pk:
    1

Specifications

About this item

The Conradson test unit is used to measure for carbon residue of petroleum products.

  • Cast iron tripod stand
  • Skidmore and spun sheet iron crucible with cover
  • Spun steel circular hood with chimney and insulation block and nichrome wire support
Certifications: ASTM D189-IP 13, D2416, BS 2000-13, FTM 791 5001, ISO 6615.