Order Entry
United Kingdom
ContactUsLinkComponent
Pyrotell®, Gel-clot formulation
Pyrotell®, Gel-clot formulation
Catalog # CAPEG5003
CAS Number:  
Pyrotell®, Gel-clot formulation
Catalog # CAPEG5003
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Pyrotell Multi test 0,03 Eu/ ml, 5 ml/vial (approx. 50 tests/vial)
  • Environmentally Preferable:
Specifications
About this item
Pyrotell® lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV's). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement. Pyrotell lysate is available in multiple sensitivities: 0,03 EU/ml or 0,25 EU/ml. For 2 and 5 ml multi-test vials, 100 µl of lysate is mixed with 100 µl of sample in a reaction tube.
  • The 2 and 5 ml vials are sold individually