SPE columns, ISOLUTE® SCX-3
Supplier: Argonaut Technologies
Certificates
About this item
ISOLUTE® SCX-3 is a ethyl benzenesulfonic acid based strong cation exchange sorbent. It is used for retention of basic compounds from aqueous samples. These sorbents manufactured using a trifunctional silane and non-endcapped. Analytes are eluted from these columns by eliminating the charge on the analyte (high pH), so this sorbent is not suitable for extraction of strong bases.
Nominal capacity of sorbent: 0,4 meq/g
Delivery: Sorbents are shipped in the protonated form.
Frequently Bought Together