BBL Enterotube II™
DIFC4373591
: DIFCO BD
:
- Pk:500 g
- Description:Enterotube Kodebog
- Environmentally Preferable:
The BD BBL Enterotube II incorporates standard biochemical tests contained in a unique, ready-to-use, compartmented tube designed to permit the simultaneous inoculation of 12 test media. This innovative product system provides rapid, accurate identification solutions with a high level of reproducibility and excellent agreement with the standard technique classically employed.
- Well-aproved
- Reliable