Syringes, Luer slip bulk, Norm-Ject®
Supplier: Air-Tite Products
New Product
Certificates
About this item
Norm-Ject® syringes are manufactured from laboratory grade PP and PE. These syringes are the choice for any situation needing an inert, non-reactive syringe.
- Contain no rubber or silicone oil or styrene or DEHP
- Latex- and DNA-free
- More chemically resistant than rubber tipped syringe
Ideal for nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures.
Frequently Bought Together