Order Entry
United Kingdom
ContactUsLinkComponent
Sigma-Aldrich® One-piece fingertight fittings
Sigma-Aldrich® One-piece fingertight fittings
Catalog # SIAL55067-U
Supplier:  Merck
Sigma-Aldrich® One-piece fingertight fittings
Catalog # SIAL55067-U
Supplier:  Merck
Specifications
  • Description:
    One-piece fittings
  • Pk:
    10
Specifications
About this item
Standard knurled PEEK fittings with 10-32 thread, no ferrule to drop onto the floor.