Anti-VEGFC Goat Polyclonal Antibody
Catalog # USBIV2110-04W
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor C
- Antigen symbol:VEGFC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:104 to 330
- Storage buffer:PBS, pH 7,2, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 0.1-0.2?g/ml; Immunochemistry: 5-15 ?g/ml
Type: Primary
Antigen: VEGFC (vascular endothelial growth factor C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human