Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SRY Mouse Monoclonal Antibody [clone: 9F639]
Anti-SRY Mouse Monoclonal Antibody [clone: 9F639]
  USBIS7049-88
Anti-SRY Mouse Monoclonal Antibody [clone: 9F639]
  USBIS7049-88

 

  • Antibody type:
    Primary
  • Antigen name:
    Sex Determining Region Y
  • Antigen symbol:
    SRY
  • Clonality:
    Monoclonal
  • Clone:
    9F639
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: SRY (sex determining region Y)
Clonality: Monoclonal
Clone: 9F639
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human