Anti-Rho-GDI beta Rabbit Polyclonal Antibody
USBIR1998-21B
- Antibody type:Primary
- Antigen symbol:Rho-GDI beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mM PBS, pH 7,4, BSA, sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: Rho-GDI beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human