Anti-PDK1 Rabbit Monoclonal Antibody [clone: 10E44]
USBIP3123-10B
- Antibody type:Primary
- Antigen name:pyruvate dehydrogenase kinase, isozyme 1
- Antigen symbol:PDK1
- Clonality:Monoclonal
- Clone:10E44
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA,
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: monoclonal
Clone: 1E+45
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat