Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-PARP 1 Rabbit Polyclonal Antibody
  USBIP3113-20Q
Anti-PARP 1 Rabbit Polyclonal Antibody
  USBIP3113-20Q

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PARP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken,
    Human,
    Bovine
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    Serum
  • Form:
    liquid
  • Amino acid number:
    197 to 214
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: ELISA: 1:100000; Immunochemistry: 1:50; Western Blot: 1:10000-1:15000

Type: Primary
Antigen: PARP 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human