Anti-NAP1L4 Rabbit Polyclonal Antibody
USBIN0023-10A-100
- Antibody type:Primary
- Antigen name:nucleosome assembly Protein 1-like 4
- Antigen symbol:NAP1L4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: NAP1L4 (nucleosome assembly protein 1-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human