Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 10D23]
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 10D23]
Catalog # USBIM9750-15J
Supplier:  US Biological
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 10D23]
Catalog # USBIM9750-15J
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mycoplasma pneumoniae
  • Clonality:
    Monoclonal
  • Clone:
    10D23
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200; Western Blot: 1:20-1:200

Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: monoclonal
Clone: 10D23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: