Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Matrix Metalloproteinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2428-99D
Supplier:  US Biological
Anti-Matrix Metalloproteinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2428-99D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 14
  • Antigen symbol:
    MMP14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    160 to 173
  • Storage buffer:
    PBS, pH 7,4, 0,2% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Matrix Metalloproteinase 14
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human