Anti-KRT9 Mouse Monoclonal Antibody [clone: Ks 9.70 and Ks 9.216]
Catalog # USBIK0199-10W
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:keratin 9
- Antigen symbol:KRT9
- Clonality:Monoclonal
- Clone:Ks 9.70 and Ks 9.216
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG3,IgG1
- Reactivity:Human
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Amino acid number:4 to 28
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:10; Immunochemistry: 1:20
Type: Primary
Antigen: KRT9 (keratin 9)
Clonality: monoclonal
Clone: Ks 9.70 and Ks 9.216
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG3
Reactivity: Human