Anti-Isoprenylcysteine Carboxymethyl Transferase Rabbit Polyclonal Antibody
Catalog # USBII9039-98
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Isoprenylcysteine Carboxymethyl Transferase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:2000
Type: Primary
Antigen: Isoprenylcysteine Carboxymethyl Transferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human