Anti-IL6R Rat Monoclonal Antibody [clone: 8H130]
Catalog # USBII8428-39M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:interleukin 6 receptor
- Antigen symbol:IL6R
- Clonality:Monoclonal
- Clone:8H130
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:20 to 357
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: IL6R (interleukin 6 receptor)
Clonality: monoclonal
Clone: 8H130
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse