Anti-Interleukin 1 Receptor Type I, p80 Rat Monoclonal Antibody [clone: 6A571]
USBII7663-25J1
- Antibody type:Primary
- Antigen name:Interleukin 1 Receptor 1
- Antigen symbol:IL1R1
- Clonality:Monoclonal
- Clone:6A571
- Conjugation:Unconjugated
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, containing 0,02% sodium azide and 0,1%BSA.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:10
Type: Primary
Antigen: Interleukin 1 Receptor Type I, p80
Clonality: monoclonal
Clone: 6A571
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse