Anti-Immunoglobulin-Like Transcript 7 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11D30]
USBII7502-76B
- Antibody type:Primary
- Antigen name:Immunoglobulin-like transcript 7
- Antigen symbol:ILT7
- Clonality:Monoclonal
- Clone:11D30
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS,up to 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: Immunoglobulin-Like Transcript 7
Clonality: monoclonal
Clone: 11D30
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human