Anti-Histone H3, trimethyl K27 Mouse monoclonal antibody [clone: 9F616]
USBIH5110-14S5
- Antibody type:Primary
- Antigen name:Histone H3
- Antigen symbol:H3
- Clonality:Monoclonal
- Clone:9F616
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Modification:trimethyl-Lys27
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:150 µg
- Pk:150 µG
Recommended dilution: ELISA: 1:1000
Type: Primary
Antigen: Histone H3, trimethyl K27
Clonality: monoclonal
Clone: 9F616
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human