Anti-Herpes Simplex Virus II, Glycoprotein G Mouse Monoclonal Antibody [clone: 5E337]
Catalog # USBIH2033-48A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Herpes Simplex Virus II, Glycoprotein G
- Clonality:Monoclonal
- Clone:5E337
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by protein A chromatography or sequential differential precipitation
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:10-1:50
Type: Primary
Antigen: Herpes Simplex Virus II, Glycoprotein G
Clonality: monoclonal
Clone: 5E337
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: