Anti-Guanine Nucleotide Binding Protein, beta 4 subunit Chicken Polyclonal Antibody
USBIG9039-31
- Antibody type:Primary
- Antigen name:Guanine Nucleotide Binding Protein, beta 4 subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 260
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:500
Type: Primary
Antigen: Guanine Nucleotide Binding Protein, beta 4 subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse